Human CLU (Clusterin) ELISA Kit
Catalog # 77511-600
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:CLU (Clusterin)
- Size:96 wells
- Sample Type:Serum, plasma, urine, saliva, tissue homogenates, cerebrospinal fluid, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:12.5 - 800 ng/ml
- Storage Temperature:2…8 °C
- Application:Apoptosis, tumor immunity, infection immunity
- Sensitivity:5.1 ng/ml
- Regulatory Status:RUO
- Cat. No.:77511-600