Rat AOPP (Advanced Oxidation Protein Products) ELISA Kit
Catalog # 77511-012
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Competitive
- Host:Rat
- Target protein:AOPP (Advanced Oxidation Protein Products)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:2 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:156.25 - 10000 pg/ml
- Storage Temperature:2…8 °C
- Application:Tumor immunity
- Sensitivity:49.7 pg/ml
- Regulatory Status:RUO
- Cat. No.:77511-012