Human 5-HT (5-Hydroxytryptamine) ELISA Kit
Catalog # 77526-202
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Competitive
- Host:Human
- Target protein:5-HT (5-Hydroxytryptamine)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates
- Time to Results:2 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:14.07 - 900 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, infection immunity, neuro science, gastroenterology
- Sensitivity:3.97 ng/ml
- Regulatory Status:RUO
- Cat. No.:77526-202