Cattle CRP (C Reactive Protein) ELISA Kit
Catalog # 77520-550
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Bovine
- Target protein:CRP (C Reactive Protein)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:6.25 - 400 ng/ml
- Storage Temperature:2…8 °C
- Application:Tumor immunity, infection immunity, cardiovascular biology, rheumatology
- Sensitivity:2.43 ng/ml
- Regulatory Status:RUO
- Cat. No.:77520-550