Mouse PDCN (Podocin) ELISA Kit
Catalog # 77519-818
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:PDCN (Podocin)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Infection immunity, kidney biomarker
- Sensitivity:0.056 ng/ml
- Regulatory Status:RUO
- Cat. No.:77519-818