Mouse CFD (Complement Factor D) ELISA Kit
77515-162
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:CFD (Complement Factor D)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.47 - 30 ng/ml
- Storage Temperature:2…8 °C
- Application:Immune molecule
- Sensitivity:0.22 ng/ml
- Regulatory Status:RUO
- Cat. No.:77515-162