Human TRH (Thyrotropin Releasing Hormone) ELISA Kit
Catalog # 77514-706
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Competitive
- Host:Human
- Target protein:TRH (Thyrotropin Releasing Hormone)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:2 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:7.82 - 500 pg/ml
- Storage Temperature:2…8 °C
- Application:Endocrinology, neuro science, hepatology, hormone metabolism
- Sensitivity:2.63 pg/ml
- Regulatory Status:RUO
- Cat. No.:77514-706