Human PDGFB (Platelet Derived Growth Factor Subunit B) ELISA Kit
Catalog # 77513-146
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:PDGFB (Platelet Derived Growth Factor Subunit B)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:15.63 - 1000 pg/ml
- Storage Temperature:2…8 °C
- Application:Cytokine
- Sensitivity:6.3 pg/ml
- Regulatory Status:RUO
- Cat. No.:77513-146