Human Lpa (Lipoprotein, a) ELISA Kit
Catalog # 77512-570
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Human
- Target protein:Lpa (Lipoprotein, a)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:3.13 - 200 ng/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway, cardiovascular biology, hepatology
- Sensitivity:1.48 ng/ml
- Regulatory Status:RUO
- Cat. No.:77512-570