To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Serum format:Frozen
- Sterility:Sterile filtered
- Description:GemCell™ Plus xeno-free, world grade human serum AB - gamma irradiated
- Size:200×100 ml
- Shipping Conditions:Ships on dry ice
- Origin:US
- Storage Temperature:Store frozen at –20...–10 ℃
- Cat. No.:77463-604
- Packaging:Bottle
Product has minimum order quantity of 200 units. GemCell™ Plus Xeno-Free, world grade is collected from a maximum of 16 healthy male donors of the AB serotype. Product is defibrinated from source plasma using therapeutic grade recombinant human thrombin. Based on extensive testing and manufacturing process that is Xeno-Free, supports clinical and commercial applications.
- This product was developed without the use of animal material
- Custom fill sizes in bottle and bag format offered
- 0.1 µm sterile-filtered
- Supports clinical and commerical applications
- Meet US and EU regulatory requirements
- Gamma irradiated