Order Entry
Puerto Rico
ContactUsLinkComponent
 
S-Chip Disposable Sperm Counting Slides
Catalog # 104016-462
Supplier:  Bulldog Bio
undefined
S-Chip Disposable Sperm Counting Slides
Catalog # 104016-462
Supplier:  Bulldog Bio
Supplier Number:  DCS-S01

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    S-Chip disposable sperm counting slides
  • Dimensions:
    25×75×2 mm
  • Size:
    50 slides/100 tests
  • Cat. No.:
    104016-462
  • No. of chambers:
    2
  • Chamber size:
    2 - 3 µl

Specifications

About this item

When it comes to in vitro fertilization (IVF) and intrauterine insemination (IUI), accurate sperm counts are vital. The S-Chip for semen testing provides exact and reliable counting while simplifying the process.

  • All-in-one disposable hemocytometer for counting sperm
  • Easy and precise counting
  • Individually sealed packaging
  • Lot tested and certified

The S-Chip contains two quartz grade optically engineered 20 µm height counting chambers that do not need coverslips. Simple operation involves pipetting the sample into each chamber, placing on the microscope, and then counting. Dispose once finished.

Packaging: 100 tests per box.