ContactUsLinkComponent
PYREX® Three-Way Connecting Tube with Parallel Sidearm and [ST] Joints, Corning
PYREX® Three-Way Connecting Tube with Parallel Sidearm and [ST] Joints, Corning
Catalog # 89091-552
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
PYREX® Three-Way Connecting Tube with Parallel Sidearm and [ST] Joints, Corning
Catalog # 89091-552
Supplier:  Corning
Supplier Number:  9040-24
Product available on GSA Advantage®
   Low Manufacturing Impact
Specifications
  • Description:
    Claisen Adapter, 160 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental Certification:
    I-RECs, ISO 14001 - Monterrey
  • Cat. No.:
    89091-552
  • Bottom [ST] joint size:
    24/40
  • Top [ST] joint size:
    [ST] 24/40
  • Supplier no.:
    9040-24
Specifications
About this item
This Claisen type three-way connecting tube features a parallel sidearm.
  • Claisen-style adapter
  • Parallel sidearm
  • All joints are [ST] 24/40
Featuring [ST] outer joints at upper end and on parallel sidearm. [ST] inner joint on lower end. All joints on each tube are [ST] 24/40.