Synthware Adapter, Gas Inlet/Outlet, Microscale, Kemtech America
60004-100
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- [ST] joint:14/10
- Tubing ext.Ø:8 mm
- Cat. No.:60004-100
- Supplier no.:A301410
Gas inlet/outlet joint adapter is suitable for microscale applications.
- For gas inlet/outlet
- [ST] joint
Adapter is equipped with a standard taper joint.