Synthware Adapters with Twin Connecting Hoses, Kemtech America
Two hose connections are placed 180° from one another on this adapter to permit a flow of inert gas over the apparatus.
- Two hose connections
- 180° between connections
Adapter features a hose size of 8 mm O.D. The hose connection is 10 mm O.D. at the largest serration.