Synthware Vacuum Adapters, Glass Stopcock, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Vacuum adapters with glass stopcoock are used to regulate the flow of gases or liquids into reaction systems.
- Inner [ST] joint
- Glass stopcock
- 90° angle hose connection
The stopcock bore is 2 mm and the hose connection O.D. is 8mm.