TechniSat® Gown Room Wiper, Texwipe®
Catalog # TW-TX1048
Supplier: Texwipe
Specifications
- Cat. No.:TW-TX1048
- W×L:12.7 x 20.3 cm (5 x 8")
- Supplier no.:TX1048
Specifications
About this item
Pre-wetted cellulose and polyester blend wiper for effective, economical wipedown of articles brought into the cleanroom.
Pre-wetted with the optimal amount of 0.2 µm-filtered, blended 6% semiconductor-grade isopropyl alcohol (IPA) and 94% deionized water solution.