SharpSafety™ In-Room™ System Wall Enclosure, Covidien
15704-170
- Color:Beige
- Dimensions:33W x 14D x 29.8H cm (13 x 51/2 x 113/4")
- Environmentally Preferable:
- Cat. No.:15704-170
- Supplier no.:85161H
Locked ABS-plastic enclosure secures a 1.9 L (2 qt.) or 4.7 L (5 qt.) sharps-disposal container for a secondary barrier of protection.
- Locked Sharps-Container Mounting
- Secure Point-of-Use Disposal
- Long-Lasting ABS Plastic Provides a Secondary Barrier of Protection
- Viewing Windows on Front and Sides
- Beige, Unobtrusive Style Fits Any Room Decor
Viewing windows on front and sides make it easy to monitor the container fill level.
This wall enclosure has a variety of convenient features. All cabinets are universally keyed, and the viewing windows on the front and sides make monitoring the fill level easy.
: Meets or exceeds JCAHO, OSHA, and NIOSH/CDC standards and requirements.
: For sharps containers, see the In-Room Sharps Disposal Systems series from Covidien.