Sharps Disposal Containers for Non-Biohazardous Waste, Covidien
Catalog # 82005-240
Supplier: Cardinal Health
Specifications
- Capacity:30.3 L (8 gal.)
- Dimensions:45.1H x 27.9W x 39.4D cm (173/4 x 11 x 151/2")
- Cat. No.:82005-240
- Supplier no.:8781
Specifications
About this item
Containers are designed to help segregate sharp waste to reduce waste costs
They are environmental green in color to differentiate them from other containers and are made with recycled plastic resin. Use them to dispose of broken light bulbs, vials, and other non-biohazardous, sharp waste
Certifications: Containers meet or exceed JCAHO, OSHA, and NIOSH/CDC standards and requirements.