Multi-Use One-Piece Sharps Collectors, BD Medical
Supplier: BD
Certificates
About this item
One-step disposal systems feature puncture-resistant polypropylene containers to prevent needle penetration
Units feature handles, tight-fitting vented lids, and safety necks so sharps cannot pop back. Translucent tops allow easy viewing of contents. Containers have sterile-indicator tape on bottom to indicate sterilization completion.
For accessories, see BD305451 series.
One-way valve guards against accidental spilling. Containers may be autoclaved and compacted, or incinerated.
Specifications
- Catalog No:
- BD305610
- BD305490
- BD305491
- BD305615
- BD305616
- BD305477
- BD305665
- BD305602
- BD305601
- BD305609
- BD305489
- BD305666
- BD305488
- Color:
- Red
- Red
- Red
- Red
- Red
- Red
- Red
- Red
- Red
- Red
- Red
- Red
- Red