Order Entry
Puerto Rico
ContactUsLinkComponent
Rubbermaid® Commercial Untouchable® Square Swing Top Lid
Rubbermaid® Commercial Untouchable® Square Swing Top Lid
  89073-716
undefined
Rubbermaid® Commercial Untouchable® Square Swing Top Lid
  89073-716
 :  RCP2664BEI

 

  • Depth:
    20⅛"
  • Color:
    Beige
  • Description:
    Square Top Lid
  • Height:
    6¼"
  • Material:
    Plastic
  • Cat. No.:
    89073-716
  • Shape:
    Square
  • For use with:
    Rubbermaid® Commercial Untouchable® 35 gal. and 50 gal. Square Containers
  • Fits:
    Container 66100-115
  • Supplier no.:
    RCP 2664 BEI
  • Lid type:
    Swing-top

 

 

Drop-and-swing design provides easy access for refuse disposal and then quickly returns to hide refuse from public view.

  • Crack-resistant
  • Controls waste, odors and smoking debris

Hygienic top for handsfree waste disposal.