About this item
Tak Pak® adhesive kits offer fast, reliable, cost-effective chemical tools for performing hand assembly operations on PCBs.
- Fast
- Reliable
- Cost-effective
Kits allow the user to easily secure change wires, mount edge guides, make adjustable components tamperproof, and relieve strain on heavy components without fear of ionic contamination.
Tak Pak® 382™ (19807-502) adhesive is an ultra-performance system with a faster fixture speed and higher viscosity for larger sized applications with a temperature range of –53 to 121°C (–65 to 250°F). It fills gaps up to 0.020cm (0.008"). Fixture time: 30sec. Full cure time: 24hrs. Viscosity: 5000cP. Kit contains a 20g (0.7oz.) bottle of adhesive and 19.8g (0.7oz.) aerosol can of 7452 accelerator.
Tak Pak® 444™ Kit (19807-500) is a standard adhesive system designed for applications with a temperature range of –53 to 82°C (–65 to 180°F). It fills gaps up to 0.018cm (0.007"). Fixture time: 30sec. Full cure time: 24hrs. Viscosity: 700cP. Kit contains a 20g (0.7oz.) bottle of adhesive and 51.8mL (1.75oz.) bottle of 7452 accelerator.