Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (APC (Allophycocyanin))
Supplier: Biotium
Certificates
About this item
CD19 (clone SJ25C1) is a validated mouse monoclonal antibody that recognizes CD19, a transmembrane glycoprotein in B cells. The antibody belongs to the Biotium Choice list of select antibodies that have been validated and optimized in-house for optimal performance.
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- 77449-122
- 77449-114
- Antigen Symbol:
- CD19
- CD19
- Antigen Synonyms:
- T-cell Surface Antigen Leu-12|CVID3|B-Lymphocytes Surface Antigen B4|B4
- T-cell Surface Antigen Leu-12|CVID3|B-Lymphocytes Surface Antigen B4|B4
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- SJ25C1
- SJ25C1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 930
- 930
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow Cytometry:
- Yes
- Yes
- Molecular Weight:
- 95 kDa
- 95 kDa
- Concentration:
- 50 µg/ml
- 50 µg/ml
- Storage Temperature:
- 2...8 °C
- 2...8 °C