Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
VeriSpec® Multi-Element Standard 23, Ricca Chemicals

VeriSpec® Multi-Element Standard 23, Ricca Chemicals

VeriSpec® Multi-Element Standard 23, 100 ml, 2% HNO₃, natural LDPE, 36 months
Catalog # RCRV010826100N
Supplier:   Ricca Chemical
Supplier #:  RV010826-100N
VeriSpec® Multi-Element Standard 23, Ricca Chemicals
Catalog # RCRV010826100N
Supplier:  Ricca Chemical
CAS Number:  
VeriSpec® Multi-Element Standard 23,  Ricca Chemicals
VeriSpec® Multi-Element Standard 23, Ricca Chemicals

VeriSpec® Multi-Element Standard 23, 100 ml, 2% HNO₃, natural LDPE, 36 months

CAS Number:  
Price
$867.08
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

15K ppm Mg, 8K ppm Na, 3.5K ppm Ca, 3000 ppm K; 100 ppb B, Fe, Mo, Sr; 50 ppb As, Ba, Ni, V, Zn

Certifications: Manufactured and Tested in an ISO 17025 accredited laboratory.

Documents

Specifications

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    natural LDPE
  • Description:
    VeriSpec® Multi-Element Standard 23
  • Size:
    100 ml
  • Shelf life:
    36 months

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.