Permabond® QFS16 Ozone-Safe Accelerator
19806-014
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- Description:113 g (4 oz.) pump spray
- Cat. No.:19806-014
- Supplier no.:QFS16-4OUNCE
This accelerator is for use with all Permabond® cyanoacrylate adhesives.
- Increases gap-filling properties and cure time
- Pump spray bottle
Accelerator is used to increase gap-filling properties and cure time.