TRUTOL® Glucose Tolerance Beverages, NERL Diagnostics
Supplier: Thermo Fisher Scientific
Certificates
About this item
Caffeine-free TRUTOL® beverages are used to screen for hyperglycemia (diabetes mellitus and gestational diabetes) and hypoglycemia (low blood sugar).
- Screening entails administering a precise dose of glucose and then testing the level of glucose in the patient’s blood at specific time intervals
- Bottles feature volume graduations imprinted on the label to aid in dispensing smaller quantities
Bottle size: 296 ml (10 oz.).
Certifications: Manufactured to meet FDA standards. Certified kosher.
Ordering information: Carbonated flavors are packaged in glass bottles. Non-carbonated flavors are packaged in plastic bottles.
Frequently Bought Together