ContactUsLinkComponent
CircuitWorks® Prototype/Repair Tools Conductive Epoxy Kit, ITW Chemtronics®
CircuitWorks® Prototype/Repair Tools Conductive Epoxy Kit, ITW Chemtronics®
Catalog # 33500-072
Supplier:  ITW Chemtronics
undefined
CircuitWorks® Prototype/Repair Tools Conductive Epoxy Kit, ITW Chemtronics®
Catalog # 33500-072
Supplier:  ITW Chemtronics
Supplier Number:  CW2400
Specifications
  • Description:
    CircuitWorks® Prototype/repair tools conductive epoxy kit
  • Cat. No.:
    33500-072
  • Supplier no.:
    CW2400
Specifications
About this item
Developed to aggresively bond to a wide variety of materials.
  • Highly moisture and chemical resistant
  • Solderless connections
  • Electrically and thermally conductive
Epoxy is highly moisture and chemical resistant and features fair flexibility. Fully cured bond has a lap shear strength of >400kg (882lbs.) and a shore hardness of >31kg (70lbs.). Volume resistivity is >0.001ohms-cm and thermal conductivity is 11.0Btu·in./(ft.²·hr.·°F). Solderless connections. Electrically and thermally conductive.
Ordering information: Kit is supplied with 14g (0.5oz.) of epoxy, reusable applicator, and instructions.