Securline® Surgical Skin Marker, Sterile, Aspen Surgical
Catalog # 52877-453
Supplier: Aspen Surgical Products
Specifications
- Description:Surgical skin marker, fine/broad tip, violet
- Environmentally Preferable:
- Cat. No.:52877-453
- Supplier no.:1002-00-PDG
Specifications
About this item
Marker features non-tattooing, surgical prep-resistant Gentian Violet ink that is nontoxic to the epidermic layer of the skin
Non-scratching. Bulk-packed.