Two Hopper Thermal Slide Printer, PiSmart
Product Details & Documents
The PiSmart slide printer incorporates two hoppers for pre-loading different slide types. Intelligent slide selection automatically determines the microscope slide required for individual samples by analyzing the label content and dispensing from the correct hopper. At 16 pounds, the PiSmart printer is light and easy to move on the benchtop.
- Industry smallest footprint
- A fast, 3 - 5 second print time
- Extremely quiet!
- Easy-to-learn interface
- Dark, high-contrast barcode printing
- Allows up to 100 characters on each slide
- Built in scanner
- LIS integration delivers a smart solution
- Four years PiSmart promise warranty
Control all of your PiSmart equipment while connecting your processes with the PathSmart tracking solution, transforming your lab to be faster, smarter, and more connected. Designed to close gaps in information, delivering additional information to support chain of custody for samples, the PathSmart hub also links all PiSmart printers to one computer in the lab, syncing user management, templates, software updates and more.
Barcode: 1D and 2D capable
Capacity: 144 slides, 72 per hopper
Documents
Specifications
- Description:Two-hopper PiSmart thermal slide printer
- Dimensions:13×6.4×16"
- Resolution:300 dpi
- Connections:Ethernet, USB-A (2), USB-B (1)
- Cat. No.:77506-768
- Weight:16 lbs.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



