ContactUsLinkComponent
Steel Hook and Bin Assortment for DuraBoard® or ¹/₈" and ¹/₄" Pegboard (24 Assorted Hooks and 2 Bins), 26-Piece
Steel Hook and Bin Assortment for DuraBoard® or ¹/₈" and ¹/₄" Pegboard (24 Assorted Hooks and 2 Bins), 26-Piece
Catalog # 77269-364
Supplier:  Triton Products
undefined
Steel Hook and Bin Assortment for DuraBoard® or ¹/₈" and ¹/₄" Pegboard (24 Assorted Hooks and 2 Bins), 26-Piece
Catalog # 77269-364
Supplier:  Triton Products
Supplier Number:  76901
Specifications
  • Color:
    Silver
  • Description:
    Steel hook and bin assortment for DuraBoard® or ¹/₈" and ¹/₄" pegboard (24 assorted hooks and 2 bins), steel/polypropylene, zinc plated, 26-piece
  • Cat. No.:
    77269-364
Specifications
About this item
DuraHook® assortments save you time, money and space. Many of the assortments contain everything you’ll need to tackle your storage challenge.
  • Unique DuraHook styles along with LocBin plastic hanging bins and BinClips can handle any pegboard storage requirement
  • Innovative double-locking system locks the hook in place with a locking tab that mounts to the pegboard from the front
  • Use on any ¹/₈" or ¹/₄" pegboard (excluding steel)
  • Hooks mount vertically and horizontally in four distinct positions allowing the use of multiple hooks for large, heavy and odd-shaped items
  • Fixtures install from the front, so no need to remove the board for installation or repositioning of DuraHooks
The DuraHook® 76901 is a 26 piece hook and hanging bin assortment kit. It contains the first double locking pegboard hooks with new and useful styles including spring clips, Extended spring clip, double loop hooks and BinClip™(s) to mount plastic hanging bins on pegboard. Hooks styles are: (3) 1" Single rod 30° bend; (2) 2¹/₂" Single rod 30° bend; (1) 6" Single rod 30° bend; (2) Pliers/wrench holder; (2) 2³/₄" Double rod 80° bend; (1) 5³/₄" Double rod 80° bend; (2) Extended spring clip; (1) Standard spring clips; (1) Double ring tool holder; (2) Curved hook; (2) 2³/₄" Double U-shape; (1) spring clip/Can holder; (1) 5" Double closed end loop; (1) Multi-prong tool holder; (2) BinClip; and (2) plastic hanging bins.
Ordering information: Limited lifetime warranty.