Orders LinkContactUsLinkComponent
DuraBoard® Pegboards, Black ABS Textured
Supplier:   Triton Products
DuraBoard® Pegboards, Black ABS Textured
Supplier:  Triton Products
Certificates
DuraBoard® Pegboards, Black ABS Textured
DuraBoard® Pegboards, Black ABS Textured
3 Options Available Below
Product Details & Documents
DuraBoard® is the most durable, weather and impact resistant pegboard available. It is eight times stronger than conventional pegboard.
  • Hole diameter is ¹/₄" with 1" O.C spacing
  • DuraBoard will never fade, dry out, crack or peel over time so it maintains holding capability over many years of use
  • Offers a professional looking pegboard system that can be cleaned without harming the board
  • Board surface is ideal for creating silhouettes of tools to be stored and managed allowing for easy tool identification
  • Use Triton 70016 mounting spacer kit to mount DuraBoard and eliminate the expense and hassle of wood framing
The commercial-grade DuraBoard® ¹/₄" textured ABS pegboard has superior holding strength with eight times that of traditional pegboard. One side has a textured finish to keep it looking new and to prevent scratching. It is the most durable, weather and impact resistant pegboard available. This pegboard will stand up to any harsh temperatures and environments and can be cleaned or washed when necessary. DuraBoard® heavy-duty pegboard is ideal for use in a clean room, factory floor production area, maintenance and repair area as well as sheds and garages. Use DuraHook® double-locking pegboard hooks to create the most secure and reliable pegboard storage system.
 : Limited lifetime warranty.
 : Mounting hardware not included.
Product Family Options
Product Information
  • Color
    Material
    Width
    Depth
    Height
    Availability
    Price
  • 77269-622
    One board, 500 lbs. (load rating per board), matte, 0.25" hole size
    One board, 500 lbs. (load rating per board), matte, 0.25" hole size
    Black
    ABS
    48"
    0.25"
    96"
    Log in for availability
    Log in for availability
    Each
    Each - $429.54
    $429.54
    Each
       $429.54
    Supplier Number: DB-96BLK
  • 77269-602
    Two boards, 250 lbs. (load rating per board), matte, 0.25" hole size
    Two boards, 250 lbs. (load rating per board), matte, 0.25" hole size
    Black
    ABS
    24"
    0.25"
    48"
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $210.41
    $210.41
    Pack of 2
       $210.41
    Supplier Number: DB-2BK
  • 77269-614
    Four boards, 250 lbs. (load rating per board), matte, 0.25" hole size
    Four boards, 250 lbs. (load rating per board), matte, 0.25" hole size
    Black
    ABS
    24"
    0.25"
    48"
    Log in for availability
    Log in for availability
    Pack of 4
    Pack of 4 - $414.14
    $414.14
    Pack of 4
       $414.14
    Supplier Number: DB-4BK
Accessories
Product Information
  • Catalog #
    Color
    Material
    Availability
    Price
  • Pegboard mounting and spacer kit for DuraBoard® or ⅛" and ¼" pegboard, 16 sets
    Pegboard mounting and spacer kit for DuraBoard® or ⅛" and ¼" pegboard, 16 sets
    77269-324
    Silver/yellow
    Steel/plastic
    Log in for availability
    Pack of 16 - $15.72