Hydrophilic Adhesive Microscope Slides, Frosted
Catalog # 76809-028
Supplier: Avantor
Specifications
- Color:White
- Description:Hydrophilic adhesive slides, frosted
- L×W:75×25 mm
- Material:Glass
- Thickness:1.0 - 1.2 mm
- Cat. No.:76809-028
Specifications
About this item
Hydrophilic adhesive slides for superior tissue adhesion and staining performance in histology labs.
- Hydrophilic surface ensures even reagent coverage, improving staining quality
- Designed for laser slide printers or manual marking
- Made of super white glass with frosted end and side
- Recommended for manual and automatic IHC staining
- Ideal for challenging tissue types in H&E staining
Featuring increased positive charges, these slides prevent tissue detachment during high-temperature processing, ensuring reliable results. The hydrophilic surface promotes uniform reagent coverage, minimizing staining issues for improved quality. Suitable for laser slide printers. Also suitable for pencil or pen marking, and slide labels.