Specifications
- Color:Silver
- Description:Zinc plated steel hook assortment for LocBoard®, 46 piece
- Cat. No.:77269-736
Specifications
About this item
A curated assortment of LocHooks® to get you up and running fast and tackle your storage and organization needs.
- Unique heavy-duty LocHook® styles can handle any pegboard storage requirement
- Hooks mount vertically and horizontally in 4 distinct positions allowing the use of multiple hooks for large, heavy and odd-shaped items
- Fixtures install from the front, so no need to remove the board for installation or repositioning of LocHooks causing no damage to the LocBoard
- Compatible with all LocBoard sizes and other brands of steel tool boards with square holes spaced 1 to ¹/₂" O.C.
Having commercial grade storage and organizational solutions is an issue that every business encounters. LocHook® and LocBoard® present an ideal steel pegboard tool storage solution that supports 5S and Lean Manufacturing initiatives. They provide the most heavy-duty pegboard and hooks system for dependable storage for factory floor, MRO, shop, home and garage. The LocHook is an innovative pegboard hook which can be mounted in any direction and is rugged enough to support just about anything. LocBoard® and LocHook® provide the perfect steel pegboard solution. Count on the power of Triton's steel pegboard system. LH1-Kit (46 pieces) includes: (5) 1" Single Rod 30 Degree Bend; (5) 4" Single Rod 30 Degree Bend; (5) 1" Double Rod 30 Degree Bend; (5) 3" Double Rod 30 Degree Bend; (5) Extended Spring Clip/Spray Can Holder; (5) Standard Spring Clip; (5) Single Ring Tool Holder; (5) 3³/₄" Curved Hook; (5) Double Closed End Loop; (1) Multi-Prong Tool Holder.