Order Entry
Puerto Rico
ContactUsLinkComponent
Human Serum AB (Off The Clot), Aseptic Assurance System
Human Serum AB (Off The Clot), Aseptic Assurance System
Catalog # 77463-594
Supplier:  GeminiBio
undefined
Human Serum AB (Off The Clot), Aseptic Assurance System
Catalog # 77463-594
Supplier:  GeminiBio
Supplier Number:  100-318-750
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Serum format:
    Frozen
  • Sterility:
    Sterile filtered
  • Description:
    Human serum AB (Off the Clot) - AAS
  • Size:
    200×100 ml
  • Shipping Conditions:
    Ships on dry ice
  • Origin:
    US
  • Storage Temperature:
    Store frozen at –20...–10 ℃
  • Cat. No.:
    77463-594
  • Packaging:
    BAGA

Specifications

About this item

Product has minimum order quantity of 200 units. Off the clot human serum AB is collected from normal healthy male donors at FDA-licensed facilities in the United States. As a natural off the clot product, this material contains no anticoagulants/preservatives. All donor units are tested for viral markers and found to be non-reactive.

  • Supports clinical and commercial applications
  • Aseptic assurance system