Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

CryoStor® CS5 Cell Freezing Medium

100 ml, Bottle
Catalog # MSPP-07933
Marketsource
 :  07933
CryoStor® CS5 Cell Freezing Medium
Catalog # MSPP-07933
Marketsource
CAS Number:  
CryoStor® CS5 Cell Freezing Medium
CryoStor® CS5 Cell Freezing Medium

100 ml, Bottle

CAS Number:  
Price
$745.38
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Animal component-free, defined cryopreservation medium with 5% DMSO.

  • Ready-to-use
  • Serum-free and protein-free
  • Animal component-free
  • cGMP manufactured with USP grade/highest-quality components
  • FDA master file
  • Sterility, endotoxin, and cell-based quality control testing

Maximize post-thaw cell recovery and viability following cryopreservation at low temperatures (−80 to −196 °C) with ready-to-use CryoStor® CS5. Serum-free, animal component-free, and cGMP-manufactured, this defined medium provides a safe, protective environment for cells and tissues during freezing and thawing processes and storage. Available in a variety of convenient formats, CryoStor® CS5 is formulated with USP-grade components and contains 5% DMSO.

Cell type: Cho Cells, Mesenchymal Stem and Progenitor Cells, Other, Pluripotent Stem Cells.

Species: Human, Mouse, Non Human Primate, Other, Rat.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Size:
    100 ml
  • Format:
    Bottle

Specifications