Simian S100A9 (S100 Calcium Binding Protein A9) ELISA Kit
Catalog # 77510-536
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Host:Monkey
- Target protein:S100A9 (S100 Calcium Binding Protein A9)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.78 - 50 ng/ml
- Storage Temperature:2…8 °C
- Application:Tumor immunity, Infection immunity, Rheumatology
- Sensitivity:0.28 ng/ml
- Regulatory Status:RUO
- Cat. No.:77510-536
Specifications
About this item
- Host: Simian (monkey)