Order Entry
Puerto Rico
ContactUsLinkComponent
Caspase-1 Substrate X, Fluorogenic, Calbiochem®
Caspase-1 Substrate X, Fluorogenic, Calbiochem®
Catalog # 80050-782
Supplier:  MilliporeSigma
CAS Number:  
Caspase-1 Substrate X, Fluorogenic, Calbiochem®
Catalog # 80050-782
Supplier:  MilliporeSigma
Supplier Number:  218739-5MG
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Caspase-1 Substrate X, Fluorogenic
  • Size:
    5 mg (0.0001 oz.)
  • Form:
    Lyophilized
  • Purity:
    95% by HPLC
  • Solubility:
    DMSO
  • Molecular Weight:
    784.8
  • Cat. No.:
    80050-782
  • Supplier no.:
    218739-5MG

Specifications

About this item

A fluorogenic substrate for detecting caspase-1 (ICE), caspase-4, and caspase-5 activity

Excitation maximum: ~365–380nm; emission maximum: ~430–460nm.