Static Shielding SECO Pack 77 Transporters, SECO
Supplier: Avantor
Certificates
About this item
Specially designed for electrostatic-free cushioning of PC boards, this cost-effective transporter pack offers triboelectric protection and cushioning from shock and vibration.
- Pouch incorporates a metallized static-shielding laminated outer layer, an amine-free static-dissipative cushion, and an amine-free dissipative inner layer
- Static charge decay: 0.05 s outer layer, 0.23 s inner layer
- Surface resistivity: 109 ohms outer layer, 1010 ohms inner layer
- Flap can be tucked in as a closure
Bags 58605-936 and 58605-938 include a foam insert.