Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-CD3E Mouse Monoclonal Antibody [clone: SK7] (RPE-Astral™616)
 :  Biotium
 
Anti-CD3E Mouse Monoclonal Antibody [clone: SK7] (RPE-Astral™616)
 :  Biotium

 

  • Catalog No:
  • 77449-060
  • 77449-058
  • Antigen Symbol:
  • CD3E
  • CD3E
  • Antigen Synonyms:
  • CD3 TCR complex|TCRE|T cell antigen receptor complex epsilon subunit of T3|T3E|T-cell surface glycoprotein CD3 epsilon chain|CD3 epsilon|TiT3 complex|T-cell surface antigen T3/Leu-4 epsilon chain
  • CD3 TCR complex|TCRE|T cell antigen receptor complex epsilon subunit of T3|T3E|T-cell surface glycoprotein CD3 epsilon chain|CD3 epsilon|TiT3 complex|T-cell surface antigen T3/Leu-4 epsilon chain
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • RPE (R-Phycoerythrin)
  • RPE (R-Phycoerythrin)
  • Clone:
  • SK7
  • SK7
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 916
  • 916
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Human thymocytes
  • Human thymocytes
  • Molecular Weight:
  • 23 kDa
  • 23 kDa
  • Concentration:
  • 100 µg/ml
  • 100 µg/ml
  • Storage Buffer:
  • PBS, 0.1% BSA, 0.05% azide
  • PBS, 0.1% BSA, 0.05% azide
  • Storage Temperature:
  • Store at 2...8 °C, protect fluorescent conjugates from light
  • Store at 2...8 °C, protect fluorescent conjugates from light
  • Shipping Temperature:
  • RT
  • RT
  • Tested Applications:
  • Flow (surface) (verified)
  • Flow (surface) (verified)

 

Frequently Bought Together