Orders LinkContactUsLinkComponent
Staticide® Diamond Polyurethane Static-Dissipative Floor Coating, ACL Staticide
Staticide® Diamond Polyurethane Static-Dissipative Floor Coating, ACL Staticide
3.8 L (1 gal.)
Catalog # 47744-784
Supplier:   ACL Staticide
Supplier #:  4700-SS 1
Staticide® Diamond Polyurethane Static-Dissipative Floor Coating, ACL Staticide
Catalog # 47744-784
Supplier:  ACL Staticide
Staticide® Diamond Polyurethane Static-Dissipative Floor Coating, ACL Staticide
Staticide® Diamond Polyurethane Static-Dissipative Floor Coating, ACL Staticide
3.8 L (1 gal.)
Price
$433.70
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
This high-performance water-based formula dissipates static discharge on concrete and other unfinished hard floor surfaces
Resists yellowing and color fading and is biodegradable.
Specifications
  • Size:
    3.8 L (1 gal.)
  • Cat. No.:
    47744-784
  • Coverage Area:
    185.8 m2 (2000 sq. ft.) per coating