Static Shield Zip-Close Bags, SECO
These clear, zip-close static-shielding bags guarantee a secure closure that can be repeatedly opened and closed.
- Low-profile, double tracked, and integrated into the bag
- Bag is reinforced with ultrasonic seals for added strength
- When zipped, a continuous static shield is formed
Static decay <1 s
Surface reistivity: <1.0×1012 Ohm inside layer, <1.0×1012 Ohm outside layer, <1.0×1012 Ohm metal layer
: The multi-layer metallized film meets or exceeds the electrical requirements of MIL-B-81705C Type III, ensuring high ESD protection.