VWR® Large Narrow Mouth Bottles, LDPE
Supplier: VWR International
Certificates
About this item
Ideal for collecting, storing, or shipping a variety of contents from distilled water to corrosive chemicals and standards.
- Leakproof
- LDPE construction
They can be stored in a freezer to −100 °C (−148 °F), and are low in trace metals, with excellent chemical resistance to most acids, bases, and alcohols. Bottles have a built-in loop on the shoulder for identification tag attachment.
Delivery: Polypropylene screw caps included.