Order Entry
Puerto Rico
ContactUsLinkComponent
 
STAT Biohazard Bag with Document Pouch
Catalog # 89005-358
Supplier:  Minigrip
undefined
STAT Biohazard Bag with Document Pouch
Catalog # 89005-358
Supplier:  Minigrip
Supplier Number:  MGTZ69S

Specifications

  • Dimensions:
    15.2×22.8 cm (6×9")
  • Thickness:
    2 mil.
  • Cat. No.:
    89005-358
  • Supplier no.:
    MGTZ69S

Specifications

About this item

Easily identifiable bag is constructed of red film and features the term STAT printed on the front.

  • TearZone® access for simple, safe use.

Bag front also features a biohazard logo and check boxes labeled Freeze, Refrigerate, and Room Temperature.

Delivery: Supplied in easy access perforated dispenser bags and can be easily ripped open for efficiency.