Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Pharmaceutical Standards, Timolol EP Impurity G
Pharmaceutical Standards, Timolol EP Impurity G
Pharmaceutical Standards, Timolol EP Impurity G
Supplier:
TLC Standards
New Product
Certificates
Expand image
Pharmaceutical Standards, Timolol EP Impurity G
Supplier:
TLC Standards
New Product
About this item
Specifications
CAS No.:
75202-36-7
Specifications