$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical Standards, Methyl 12-Ketostearate
Pharmaceutical Standards, Methyl 12-Ketostearate
:
TLC Standards
New Product
Pharmaceutical Standards, Methyl 12-Ketostearate
:
TLC Standards
New Product
CAS No.:
2380-27-0