$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical Standards, Amisulpride EP Impurity F (Amisulpride N-Oxide)
Pharmaceutical Standards, Amisulpride EP Impurity F (Amisulpride N-Oxide)
Pharmaceutical Standards, Amisulpride EP Impurity F (Amisulpride N-Oxide)
Supplier:
TLC Standards
New Product
Certificates
Expand image
Pharmaceutical Standards, Amisulpride EP Impurity F (Amisulpride N-Oxide)
Supplier:
TLC Standards
New Product
About this item
Specifications
CAS No.:
71676-01-2
Specifications