$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical Standards, Atosiban Acetate
Pharmaceutical Standards, Atosiban Acetate
Pharmaceutical Standards, Atosiban Acetate
Supplier:
TLC Standards
New Product
Certificates
Expand image
Pharmaceutical Standards, Atosiban Acetate
Supplier:
TLC Standards
New Product
About this item
Specifications
CAS No.:
914453-95-5
Specifications