$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical Standards, Mesalazine (Mesalamine) EP Impurity I
Pharmaceutical Standards, Mesalazine (Mesalamine) EP Impurity I
:
TLC Standards
New Product
Pharmaceutical Standards, Mesalazine (Mesalamine) EP Impurity I
:
TLC Standards
New Product
CAS No.:
3147-53-3