ContactUsLinkComponent
VWR® Protection Hood and Face Cover
VWR® Protection Hood and Face Cover
Catalog # 76292-068
Supplier:  VWR International
undefined
VWR® Protection Hood and Face Cover
Catalog # 76292-068
Supplier:  VWR International
Specifications
  • Clothing Size:
    XL
  • Color:
    White
  • Cat. No.:
    76292-068
Specifications
About this item
The lightweight VWR® protection hood and face cover provides a comfortable fit and meets quality gowning standards.
  • Lightweight hood and face combination
  • Comfortable fit
  • SPP 20 g
  • Latex-free
The VWR® protection hood and face cover protects users from potential environmental contaminations.
Packaging: Hoods are packed 100 per bag, 5 bags per case.