Order Entry
Puerto Rico
ContactUsLinkComponent
Polyethylene glycol 1,450, waxy soft solid, J.T.Baker®
Polyethylene glycol 1,450, waxy soft solid, J.T.Baker®
Catalog # JTU220-7
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Polyethylene glycol 1,450, waxy soft solid, J.T.Baker®
Catalog # JTU220-7
Supplier Number:  U220-07
CAS Number:  
MOC:  Non-RegulatedCertificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Size:
    500 ml
  • Cat. No.:
    JTU220-7
  • Supplier no.:
    U220-07

Specifications

Specification Test Results

  • Appearance:
    Passes Test
  • Identification (by IR):
    Passes Test

Specification Test Results

About this item

 :
 :1450 g/mol
 :Management of Change (MOC) category = N
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839