


Tetrahydrofuran ≥99.5% (by GC, corrected for water content) stabilized for HPLC, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIALS US
MOC: Non-Regulated
About this item
CAS Number:109-99-9
:
Formula:C₄H₈O
MDL Number:MFCD00005356
:72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:–21.5 °C
UN:2056
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 75 - 400 ppm
Storage Temperature:Ambient
EINECS:203-726-8
REACH:05-2114615188-48
:GHS02
:GHS08
:GHS07
Specification Test Results
- Assay (by GC, corrected for water, exclusive of preservative):≥99.5 %
- Color (APHA):≤10
- Peroxide (as H₂O₂):≤0.005 %
- Stabilizer (BHT):75 - 400 ppm
- Water (by KF, coulometric):≤0.010 %
Specification Test Results
Related Information
Certificates
Search by Lot Number9440 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor High-Purity Solvents and Reagents Avantor J.T.Baker® LCMS and HPLC Solvents and Reagents Avantor Portfolio of Chemicals Avantor Top Rated Guide
Product Literature
JTBaker 9440 Datasheet